Assay Details
Target Gene Details
Entrez Gene ID: | 23872 |
Gene Name: | E26 avian leukemia oncogene 2, 3' domain |
Gene Aliases: |
AU022856, Ets-2 |
Location: |
Chr.16:95702407-95721049 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ets2 | NM_011809.3 | NP_035939.3 | ||
| AK014602.2 | ||||
| AK038348.1 | BAE20535.1 | |||
| AK128933.1 | ||||
| AK143687.1 | BAE25498.1 | |||
| AK144974.1 | BAE26161.1 | |||
| AK145852.1 | BAE26698.1 | |||
| AK150175.1 | ||||
| AK151315.1 | BAE30296.1 | |||
| AK152750.1 | ||||
| AK153339.1 | BAE31917.1 | |||
| AK153478.1 | BAE32028.1 | |||
| BC005486.1 | AAH05486.1 | |||
| BC005504.1 | AAH05504.1 | |||
| CT010235.1 | CAJ18443.1 | |||
| J04103.1 | AAA37581.1 |
More Information
Set Membership: |
|

Genomic Map