Assay Details
Target Gene Details
Entrez Gene ID: | 11820 |
Gene Name: | amyloid beta (A4) precursor protein |
Gene Aliases: |
Abeta, Abpp, Adap, Ag, Cvap, E030013M08Rik, betaApp |
Location: |
Chr.16:84954436-85173952 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 16 - Exon 16 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
App | NM_001198823.1 | NP_001185752.1 | ||
NM_001198824.1 | NP_001185753.1 | |||
NM_001198825.1 | NP_001185754.1 | |||
NM_001198826.1 | NP_001185755.1 | |||
NM_007471.3 | NP_031497.2 | |||
AK052448.1 | BAC34997.1 | |||
AK076506.1 | BAC36369.1 | |||
AK128971.1 | ||||
AK133639.1 | ||||
AK147543.1 | BAE27986.1 | |||
AK148439.1 | ||||
AK159245.1 | BAE34927.1 | |||
AK159718.1 | BAE35313.1 | |||
AK161854.1 | ||||
AK161976.1 | ||||
AK190051.1 | ||||
AK191213.1 | ||||
AK191703.1 | ||||
AK191711.1 | ||||
AK192103.1 | ||||
AK192320.1 | ||||
AK192899.1 | ||||
AK192938.1 | ||||
AK194133.1 | ||||
AK197763.1 | ||||
AK198568.1 | ||||
AK199923.1 | ||||
AK200960.1 | ||||
AK202486.1 | ||||
AK202618.1 | ||||
AK216049.1 | ||||
AK216591.1 | ||||
AY267348.1 | AAP23169.1 | |||
BC005490.1 | AAH05490.1 | |||
BC070409.1 | AAH70409.1 | |||
M18373.1 | AAA37139.1 | |||
U84012.1 | AAB41502.1 | |||
X59379.1 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
Gene Ontology Categories:
