Assay Details
Target Gene Details
Entrez Gene ID: | 12028 |
Gene Name: | BCL2-associated X protein |
Gene Aliases: |
- |
Location: |
Chr.7:45461695-45466903 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bax | NM_007527.3 | NP_031553.1 | ||
| AK013298.1 | BAB28776.1 | |||
| AK149994.1 | BAE29222.1 | |||
| AK159233.1 | BAE34919.1 | |||
| AY095934.1 | AAM34436.1 | |||
| BC018228.1 | AAH18228.1 | |||
| BC053380.1 | AAH53380.1 | |||
| L22472.1 | AAA03622.1 |
More Information
Additional Information:
Set Membership: |
|

Genomic Map