Assay Details
Target Gene Details
Entrez Gene ID: | 74591 |
Gene Name: | ATP-binding cassette, sub-family A (ABC1), member 12 |
Gene Aliases: |
4832428G11Rik, 4833417A11Rik |
Location: |
Chr.1:71243090-71414910 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 40 - Exon 40 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abca12 | NM_175210.3 | NP_780419.2 | ||
| BC158053.1 | ||||
| BC158063.1 |
More Information
Set Membership: |
|

Genomic Map