Assay Details
Target Gene Details
Entrez Gene ID: | 11352 |
Gene Name: | v-abl Abelson murine leukemia viral oncogene 2 (arg, Abelson-related gene) |
Gene Aliases: |
AA536808, Abll, Arg |
Location: |
Chr.1:156558171-156649619 on Build GRCm38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abl2 | NM_001136104.1 | NP_001129576.1 | ||
| NM_009595.3 | NP_033725.2 | |||
| BC137771.1 | ||||
| BC144897.1 | ||||
| DQ084361.1 | AAY86039.1 |
More Information
Set Membership: |
|

Genomic Map