Assay Details
Target Gene Details
Entrez Gene ID: | 71724 |
Gene Name: | aldehyde oxidase 3 |
Gene Aliases: |
1200011D03Rik, AI326299, Aoh1, Aoh2 |
Location: |
Chr.1:58113136-58200452 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 23 - Intron 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aox3 | NM_023617.2 | NP_076106.2 | ||
| AF172276.2 | AAD51028.2 | |||
| AK004700.1 | BAB23485.1 | |||
| AK030109.1 | ||||
| AK149434.1 | ||||
| AY347569.1 | AAQ24537.1 | |||
| AY347570.1 | AAQ24538.1 | |||
| BC138876.1 |

Genomic Map