Assay Details
Target Gene Details
Entrez Gene ID: | 16764 |
Gene Name: | AF4/FMR2 family, member 3 |
Gene Aliases: |
3222402O04Rik, A730046J16, LAF-4, Laf4 |
Location: |
Chr.1:38175991-38627837 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aff3 | NM_001290814.1 | NP_001277743.1 | ||
| AK014322.1 |
More Information
Set Membership: |
|

Genomic Map