Assay Details
Target Gene Details
Entrez Gene ID: | 72949 |
Gene Name: | cyclin T2 |
Gene Aliases: |
2900041I18Rik, C81304, CycT2, CycT2a, CycT2b |
Location: |
Chr.1:127774164-127804837 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ccnt2 | NM_028399.1 | NP_082675.1 | ||
| AK047125.1 | ||||
| AK049863.1 | ||||
| AK152399.1 | BAE31187.1 | |||
| AK154439.1 | BAE32588.1 | |||
| BC054122.1 | AAH54122.1 | |||
| JX965957.1 | ||||
| JX965958.1 |

Genomic Map