Assay Details
Target Gene Details
Entrez Gene ID: | 67426 |
Gene Name: | aarF domain containing kinase 3 |
Gene Aliases: |
4632432J16Rik, AI462003, Cabc1, mKIAA0451 |
Location: |
Chr.1:180165238-180196020 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Intron 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adck3 | NM_001163290.1 | NP_001156762.1 | ||
| NM_023341.3 | NP_075830.2 | |||
| AJ278735.1 | CAC00538.1 | |||
| AK004177.1 | ||||
| AK004791.1 | BAB23567.2 | |||
| AK014605.2 | BAB29459.2 | |||
| AK186293.1 | ||||
| AK191635.1 | ||||
| AK192575.1 | ||||
| AK194890.1 | ||||
| AK196005.1 | ||||
| AK200253.1 | ||||
| AK202430.1 | ||||
| AK207786.1 | ||||
| AK217974.1 | ||||
| BC030937.1 | AAH30937.1 | |||
| U31629.1 | AAA86413.1 |
More Information
Set Membership: |
|

Genomic Map