Assay Details
Target Gene Details
Entrez Gene ID: | 67771 |
Gene Name: | actin related protein 2/3 complex, subunit 5 |
Gene Aliases: |
5830443F10Rik, p16-Arc |
Location: |
Chr.1:152766542-152775580 on Build GRCm38 |
Assay Gene Location: | Within Intron 1 |
More Information
Set Membership: |
|

Genomic Map