Assay Details
Target Gene Details
Entrez Gene ID: | 12633 |
Gene Name: | CASP8 and FADD-like apoptosis regulator |
Gene Aliases: |
2310024N18Rik, A430105C05Rik, CLARP, Cash, Casper, ENSMUSG00000072980, FLAME, FLAME-1, Flip, Gm9845, I-FLICE, MRIT, c-Flip |
Location: |
Chr.1:58711310-58759209 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cflar | NM_001289704.2 | NP_001276633.1 | ||
| NM_001293804.1 | NP_001280733.1 | |||
| NM_001293805.1 | NP_001280734.1 | |||
| NM_009805.4 | NP_033935.2 | |||
| NM_207653.5 | NP_997536.1 | |||
| AK143812.1 | BAE25546.1 | |||
| AK152515.1 | ||||
| AK161120.1 | ||||
| BC029223.1 | AAH29223.1 | |||
| BC085105.1 | AAH85105.1 | |||
| U97076.1 | AAC53281.1 | |||
| Y14041.1 | CAA74368.1 | |||
| Y14042.1 | CAA74369.1 |
More Information
Set Membership: |
|

Genomic Map