Assay Details
Target Gene Details
Entrez Gene ID: | 13869 |
Gene Name: | v-erb-a erythroblastic leukemia viral oncogene homolog 4 (avian) |
Gene Aliases: |
Her4, c-erbB-4 |
Location: |
Chr.1:68031904-69108794 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Erbb4 | NM_010154.1 | NP_034284.1 | ||
| AK144050.1 | BAE25671.1 |
More Information
Set Membership: |
|

Genomic Map