Assay Details
Target Gene Details
Entrez Gene ID: | 227331 |
Gene Name: | GRB10 interacting GYF protein 2 |
Gene Aliases: |
2610016F01Rik, A830080H02Rik, AI852361, AW259676, BC006835, Tnrc15, mKIAA0642 |
Location: |
Chr.1:87326981-87450810 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gigyf2 | NM_001110212.2 | 1 | 60 | NP_001103682.1 |
| NM_146112.4 | 1 | 60 | NP_666224.3 | |
| AK011414.1 | ||||
| AK034426.1 | ||||
| AK079246.1 | BAC37586.1 | |||
| AK137375.1 | BAE23332.1 | |||
| AK144058.1 | BAE25675.1 | |||
| AK160878.1 | BAE36066.1 | |||
| AY176043.1 | AAO46887.1 | |||
| BC089036.1 | ||||
| BY117987.1 | 1 | 60 |

Genomic Map