Assay Details
Target Gene Details
Entrez Gene ID: | 381314 |
Gene Name: | isoleucine-tRNA synthetase 2, mitochondrial |
Gene Aliases: |
2010002H18Rik, C79125 |
Location: |
Chr.1:185286662-185329419 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Iars2 | NM_198653.2 | NP_941055.1 | ||
| AK036182.1 | BAC29337.1 | |||
| AK043590.1 | ||||
| AK043729.1 | BAC31631.1 | |||
| AK048653.1 | BAC33410.1 | |||
| AK050358.1 | BAC34208.1 | |||
| AK051778.1 | ||||
| AK078183.1 | ||||
| AK085714.1 | ||||
| AK088965.1 | ||||
| AK164435.1 | 4 | 1225 | ||
| AK166832.1 | BAE39055.1 | |||
| BC052403.1 | AAH52403.1 |
More Information
Set Membership: |
|

Genomic Map