Assay Details
Target Gene Details
Entrez Gene ID: | 55927 |
Gene Name: | hairy and enhancer of split 6 |
Gene Aliases: |
AI326893, bHLHb41 |
Location: |
Chr.1:91411482-91413877 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hes6 | NM_019479.3 | NP_062352.1 | ||
| AB035178.1 | BAA96081.1 | |||
| AF247040.1 | AAF63757.1 | |||
| AF260236.1 | AAK51633.1 | |||
| AK005022.1 | ||||
| AK007480.1 | ||||
| AK012582.1 | 3 | 306 | ||
| AK013443.1 | BAB28856.1 | |||
| AK076135.1 | BAC36210.1 | |||
| AK146192.1 | 3 | 267 | ||
| AK155340.1 | BAE33204.1 | |||
| BC012897.1 | AAH12897.1 |
More Information
Set Membership: |
|

Genomic Map