Assay Details
Target Gene Details
Entrez Gene ID: | 227331 |
Gene Name: | GRB10 interacting GYF protein 2 |
Gene Aliases: |
2610016F01Rik, A830080H02Rik, AI852361, AW259676, BC006835, Tnrc15, mKIAA0642 |
Location: |
Chr.1:87326981-87450810 on Build GRCm38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gigyf2 | NM_001110212.2 | 19 | 2342 | NP_001103682.1 |
| NM_146112.4 | 19 | 2360 | NP_666224.3 | |
| AK079246.1 | 19 | 2312 | BAC37586.1 | |
| AK122337.1 | 10 | 1431 | BAC65619.1 | |
| AK144058.1 | 19 | 2322 | BAE25675.1 | |
| AK160878.1 | 17 | 2154 | BAE36066.1 | |
| AY176043.1 | 19 | 2306 | AAO46887.1 | |
| BC089036.1 | 19 | 2316 |

Genomic Map