Assay Details
Target Gene Details
Entrez Gene ID: | 212442 |
Gene Name: | lactamase, beta 2 |
Gene Aliases: |
Cgi-83, E430032H21Rik |
Location: |
Chr.1:13625893-13660605 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lactb2 | NM_145381.2 | NP_663356.1 | ||
| AK088932.1 | BAC40659.1 | |||
| AK090091.1 | ||||
| AK143912.1 | BAE25598.1 | |||
| BC004045.1 | AAH04045.1 |
More Information
Set Membership: |
|

Genomic Map