Assay Details
Target Gene Details
Entrez Gene ID: | 16782 |
Gene Name: | laminin, gamma 2 |
Gene Aliases: |
AA589349 |
Location: |
Chr.1:153122756-153186447 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lamc2 | NM_008485.3 | NP_032511.3 | ||
| BB617855.1 | ||||
| L20477.1 | AAA68093.1 | |||
| U43327.2 | AAA85256.2 |
More Information
Set Membership: |
|

Genomic Map