Assay Details
Target Gene Details
Entrez Gene ID: | 329252 |
Gene Name: | leucine-rich repeat-containing G protein-coupled receptor 6 |
Gene Aliases: |
A530037C04Rik, D830026M09 |
Location: |
Chr.1:134983300-135105374 on Build GRCm38 |
Assay Gene Location: | Within Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lgr6 | NM_001033409.3 | NP_001028581.1 | ||
| AK052873.1 | ||||
| AK085901.1 | ||||
| AK137380.1 | BAE23335.1 | |||
| BC141210.1 |
More Information
Set Membership: |
|

Genomic Map