Assay Details
Target Gene Details
Entrez Gene ID: | 16865 |
Gene Name: | eukaryotic translation initiation factor 2D |
Gene Aliases: |
D1Ertd5e, Lgtn |
Location: |
Chr.1:131153170-131187659 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 11 - Intron 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Eif2d | NM_001136070.1 | NP_001129542.1 | ||
| NM_010709.3 | NP_034839.2 | |||
| AK030312.1 | ||||
| AK034115.1 | BAC28591.1 | |||
| AK036154.1 | BAC29323.1 | |||
| AK046539.1 | BAC32777.1 | |||
| AK047005.1 | BAC32939.1 | |||
| AK082748.1 | BAC38598.1 | |||
| AK083638.1 | BAC38976.1 | |||
| AK139052.1 | BAE23874.1 | |||
| AK161269.1 | BAE36281.1 | |||
| AK166085.1 | ||||
| AK167980.1 | BAE39972.1 | |||
| AK168120.1 | BAE40091.1 | |||
| AK170252.1 | BAE41662.1 | |||
| AK170448.1 | BAE41806.1 | |||
| AK171115.1 | BAE42257.1 | |||
| BC024501.1 | AAH24501.1 | |||
| BC025036.1 | AAH25036.1 | |||
| U58337.1 | AAC53056.1 |
More Information
Set Membership: |
|

Genomic Map