Assay Details
Target Gene Details
Entrez Gene ID: | 68421 |
Gene Name: | LMBR1 domain containing 1 |
Gene Aliases: |
0910001K20Rik, AV347960 |
Location: |
Chr.1:24678544-24766301 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lmbrd1 | NM_026719.2 | NP_080995.2 | ||
| AK152170.1 | BAE31002.1 | |||
| AK153237.1 | BAE31829.1 | |||
| AK158982.1 | BAE34756.1 | |||
| AK167080.1 | BAE39240.1 | |||
| AY061989.1 | AAL38052.1 | |||
| BC025579.1 | AAH25579.1 | |||
| BC027774.1 | AAH27774.1 | |||
| BC031902.1 | AAH31902.1 | |||
| BC039053.1 | AAH39053.1 |
More Information
Set Membership: |
|

Genomic Map