Assay Details
Target Gene Details
Entrez Gene ID: | 107734 |
Gene Name: | mitochondrial ribosomal protein L30 |
Gene Aliases: |
2310001L22Rik, MRP-L28, Rpml28 |
Location: |
Chr.1:37890498-37898338 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mrpl30 | NM_027098.2 | NP_081374.1 | ||
| AK009074.1 | BAB26057.1 | |||
| AK085466.1 | ||||
| AK089015.1 | BAC40703.1 |
More Information
Set Membership: |
|

Genomic Map