Assay Details
Target Gene Details
Entrez Gene ID: | 100380944 |
Gene Name: | INO80 complex subunit D, opposite strand |
Gene Aliases: |
Gm11602, OTTMUSG00000002611 |
Location: |
Chr.1:63114471-63158295 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ino80dos | NR_045914.1 | |||
NR_045915.1 | ||||
AK017924.1 |
Target Gene Details
Entrez Gene ID: | 227197 |
Gene Name: | NADH dehydrogenase (ubiquinone) Fe-S protein 1 |
Gene Aliases: |
5830412M15Rik, 9930026A05Rik |
Location: |
Chr.1:63143592-63176822 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 18 - Intron 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ndufs1 | NM_001160038.1 | NP_001153510.1 | ||
NM_001160039.1 | NP_001153511.1 | |||
NM_001160040.1 | NP_001153512.1 | |||
NM_145518.2 | NP_663493.2 | |||
AK034597.1 | BAE20494.1 | |||
AK036926.1 | BAC29641.1 | |||
AK142711.1 | BAE25170.1 | |||
AK146814.1 | BAE27454.1 | |||
AK167705.1 | BAE39749.1 | |||
BC006660.1 | AAH06660.1 | |||
BC015300.1 | AAH15300.1 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
