Assay Details
Target Gene Details
Entrez Gene ID: | 13924 |
Gene Name: | protein tyrosine phosphatase, receptor type, V |
Gene Aliases: |
Esp, OST, OST-PTP, mOST-PTP |
Location: |
Chr.1:135108498-135132575 on Build GRCm38 |
Assay Gene Location: | Within Exon 31 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ptprv | NM_007955.3 | 31 | 5229 | NP_031981.3 |
| AF300701.1 | 30 | 4706 | AAG28768.1 | |
| AK156112.1 | 31 | 5232 | ||
| U36488.1 | 27 | 4391 | AAC52868.1 |
More Information
Set Membership: |
|

Genomic Map