Assay Details
Target Gene Details
Entrez Gene ID: | 109294 |
Gene Name: | phosphatidylinositol-3,4,5-trisphosphate-dependent Rac exchange factor 2 |
Gene Aliases: |
6230420N16Rik, AI316880, AI553603, C030045D06Rik, D430013K02, Depdc2, P-Rex2 |
Location: |
Chr.1:10993273-11303683 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 17 - Intron 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prex2 | NM_029525.1 | NP_083801.1 | ||
| AK018093.1 | BAB31066.1 | |||
| AK138884.1 | BAE23811.1 | |||
| AK142414.1 | BAE25059.1 | |||
| AM109952.1 | CAJ33348.1 |
More Information
Set Membership: |
|

Genomic Map