Assay Details
Target Gene Details
Entrez Gene ID: | 29809 |
Gene Name: | RAB GTPase activating protein 1-like |
Gene Aliases: |
5830411O09Rik, 8430421H08Rik, 9630005B12Rik, AW049894, HHL, Hh1, mKIAA0471 |
Location: |
Chr.1:160219174-160793476 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 20 - Intron 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Rabgap1l | NM_001038621.2 | NP_001033710.1 | ||
| NM_013862.5 | NP_038890.3 | |||
| AK020007.1 | ||||
| AK035796.1 | BAC29189.1 | |||
| AK039993.1 | ||||
| AK079402.1 | ||||
| AK088910.1 | ||||
| AK122292.1 | BAC65574.1 | |||
| AK147634.1 | BAE28037.1 | |||
| BC038651.1 | AAH38651.1 | |||
| BC145811.1 | ||||
| BC145813.1 |
More Information
Set Membership: |
|

Genomic Map