Assay Details
Target Gene Details
Entrez Gene ID: | 20343 |
Gene Name: | selectin, lymphocyte |
Gene Aliases: |
AI528707, CD62L, L-selectin, LECAM-1, Lnhr, Ly-22, Ly-m22, Lyam-1, Lyam1 |
Location: |
Chr.1:164061813-164080785 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Sell | NM_001164059.1 | NP_001157531.1 | ||
NM_011346.2 | NP_035476.1 | |||
AB353767.1 | ||||
AB353768.1 | ||||
AK137509.1 | BAE23389.1 | |||
AK157273.1 | ||||
AK161754.1 | BAE36559.1 | |||
AK163066.1 | BAE37180.1 | |||
AK170753.1 | BAE42004.1 | |||
AK172116.1 | BAE42834.1 | |||
BC052681.1 | AAH52681.1 | |||
M25324.1 | AAA39431.1 | |||
M36005.1 | AAA39722.1 | |||
M36058.1 | AAA39723.1 | |||
X14772.1 | CAA32880.1 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
