Assay Details
Target Gene Details
Entrez Gene ID: | 54380 |
Gene Name: | SWI/SNF related matrix associated, actin dependent regulator of chromatin, subfamily a-like 1 |
Gene Aliases: |
6030401P21Rik, AI851716, Mharp |
Location: |
Chr.1:72536687-72636790 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 2 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smarcal1 | NM_018817.2 | NP_061287.2 | ||
| AF088884.1 | AAF24985.1 | |||
| AK035854.1 | ||||
| AK042332.1 | BAE20630.1 | |||
| AK077878.1 | BAC37047.1 | |||
| AK083488.1 | BAC38933.1 | |||
| AK146767.1 | ||||
| AK154020.1 | BAE32320.1 | |||
| AK161570.1 | ||||
| AK169465.1 | BAE41189.1 | |||
| BC029078.1 | AAH29078.1 |
More Information
Set Membership: |
|

Genomic Map