Assay Details
Target Gene Details
Entrez Gene ID: | 74150 |
Gene Name: | solute carrier family 35, member F5 |
Gene Aliases: |
1300003P13Rik, AI646727 |
Location: |
Chr.1:125560414-125595780 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc35f5 | NM_028787.4 | NP_083063.2 | ||
| AK004892.2 | BAB23648.1 | |||
| AK076339.1 | ||||
| AK145848.1 | BAE26695.1 | |||
| AK169012.1 | BAE40809.1 | |||
| AK171951.1 | BAE42744.1 | |||
| BC026858.1 | AAH26858.1 |
More Information
Set Membership: |
|

Genomic Map