Assay Details
Target Gene Details
Entrez Gene ID: | 81879 |
Gene Name: | transcription factor CP2-like 1 |
Gene Aliases: |
1810030F05Rik, 4932442M07Rik, AA575098, Cp2l1, Crtr-1, D930018N21Rik, LBP-9, Tcfcp2l1 |
Location: |
Chr.1:118627923-118685168 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 8 - Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tfcp2l1 | NM_023755.2 | NP_076244.2 | ||
| AF311309.1 | AAK01150.1 | |||
| AK143960.1 | BAE25632.1 | |||
| BC116663.1 |
More Information
Set Membership: |
|

Genomic Map