Assay Details
Target Gene Details
Entrez Gene ID: | 21749 |
Gene Name: | telomeric repeat binding factor 1 |
Gene Aliases: |
Pin2, Trbf1, Trf1 |
Location: |
Chr.1:15804982-15844052 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Terf1 | NM_001286628.1 | NP_001273557.1 | ||
NM_009352.3 | NP_033378.1 | |||
AB109563.1 | BAC76782.1 | |||
AK132022.1 | BAE20947.1 | |||
BC034866.1 | AAH34866.1 | |||
U65586.1 | AAB53970.1 |
