Assay Details
Target Gene Details
Entrez Gene ID: | 208263 |
Gene Name: | torsin A interacting protein 1 |
Gene Aliases: |
LAP1, Lap1B |
Location: |
Chr.1:156004599-156036480 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Intron 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tor1aip1 | NM_001160018.1 | NP_001153490.1 | ||
| NM_001160019.1 | NP_001153491.1 | |||
| NM_144791.2 | NP_659040.2 | |||
| AB251963.1 | ||||
| AK149727.1 | BAE29050.1 | |||
| AK152751.1 | BAE31466.1 | |||
| BC010841.1 | AAH10841.1 | |||
| BQ891636.1 | ||||
| CA465752.1 |
More Information
Set Membership: |
|

Genomic Map