Assay Details
Target Gene Details
Entrez Gene ID: | 56207 |
Gene Name: | ubiquitin carboxyl-terminal esterase L5 |
Gene Aliases: |
5830413B11Rik, Uch37 |
Location: |
Chr.1:143777221-143807466 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Uchl5 | NM_001159866.1 | NP_001153338.1 | ||
| NM_019562.2 | NP_062508.2 | |||
| AF148447.1 | AAD31534.1 | |||
| AF175903.1 | AAD50311.1 | |||
| AK007860.1 | BAB25312.1 | |||
| AK011117.1 | BAB27412.1 | |||
| AK017925.1 | BAB31005.1 | |||
| BC006891.1 | AAH06891.1 |
More Information
Set Membership: |
|

Genomic Map