Assay Details
Target Gene Details
Entrez Gene ID: | 75472 |
Gene Name: | cilia and flagella associated protein 126 |
Gene Aliases: |
1700009P17Rik, AV282292, Flattop, Fltp |
Location: |
Chr.1:171113744-171126967 on Build GRCm38 |
Assay Gene Location: | Within Exon 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Cfap126 | NM_001081275.1 | 5 | 532 | NP_001074744.1 |
AK005804.1 | 5 | 534 | BAB24245.1 | |
BC061017.1 | 4 | 258 | AAH61017.1 |
