Assay Details
Target Gene Details
Entrez Gene ID: | 66359 |
Gene Name: | COX20 Cox2 chaperone |
Gene Aliases: |
2310005N03Rik, Fam36a |
Location: |
Chr.1:178319153-178322693 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cox20 | NM_025511.2 | NP_079787.1 | ||
| AK009178.1 | BAB26124.1 | |||
| BC055277.1 | AAH55277.1 | |||
| BC059717.1 | AAH59717.1 | |||
| BY779933.1 |
Target Gene Details
Entrez Gene ID: | 51810 |
Gene Name: | heterogeneous nuclear ribonucleoprotein U |
Gene Aliases: |
AA408410, AI256620, AL024194, AL024437, AW557595, C86794, Hnrpu, SAFA, Sp120, hnRNP U |
Location: |
Chr.1:178321108-178337810 on Build GRCm38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Hnrnpu |
More Information
Set Membership: |
|

Genomic Map