Assay Details
Target Gene Details
Entrez Gene ID: | 68691 |
Gene Name: | KAT8 regulatory NSL complex subunit 1-like |
Gene Aliases: |
1110028C15Rik, 1190030G24, AI427598, C430010P07Rik, mKIAA4189 |
Location: |
Chr.1:66719243-66817596 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kansl1l | NM_001122738.1 | 8 | 2115 | NP_001116210.1 |
| NM_177645.4 | 8 | 2220 | NP_808313.3 | |
| AK082897.1 | 7 | 935 | BAC38675.1 | |
| AK145440.1 | 8 | 2192 | ||
| AK220534.1 | 8 | 2171 | BAD90312.1 | |
| BC141036.1 | 9 | 2130 |

Genomic Map