Assay Details
Target Gene Details
Entrez Gene ID: | 215243 |
Gene Name: | TRAF3 interacting protein 3 |
Gene Aliases: |
6030423D04Rik, AI115021, T3jam |
Location: |
Chr.1:193175504-193201546 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Traf3ip3 | NM_153137.4 | NP_694777.3 | ||
| AK031400.1 | BAC27388.1 | |||
| AK161211.1 | ||||
| AY383616.1 | AAQ84304.1 | |||
| BC137605.1 | ||||
| BC171933.1 |
More Information
Set Membership: |
|

Genomic Map