Assay Details
Target Gene Details
Entrez Gene ID: | 74104 |
Gene Name: | ATP-binding cassette, sub-family B (MDR/TAP), member 6 |
Gene Aliases: |
1200005B17Rik |
Location: |
Chr.1:75171640-75180283 on Build GRCm38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abcb6 | NM_023732.2 | 15 | 2262 | NP_076221.1 |
| AK004605.1 | 15 | 2342 | BAB23404.1 | |
| AK040323.1 | 10 | 3509 | ||
| AK151165.1 | 15 | 2371 | BAE30168.1 | |
| AK164003.1 | 13 | 3057 | ||
| AK168642.1 | 15 | 2363 | BAE40501.1 | |
| AK200467.1 | ||||
| AK200865.1 | ||||
| AK208919.1 | ||||
| AK210369.1 | ||||
| AK212610.1 | ||||
| AK218549.1 | ||||
| BC006634.1 | 15 | 2262 | AAH06634.1 |
More Information
Set Membership: |
|

Genomic Map