Assay Details
Target Gene Details
Entrez Gene ID: | 11352 |
Gene Name: | v-abl Abelson murine leukemia viral oncogene 2 (arg, Abelson-related gene) |
Gene Aliases: |
AA536808, Abll, Arg |
Location: |
Chr.1:156558171-156649619 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Abl2 | NM_001136104.1 | 3 | 522 | NP_001129576.1 |
NM_009595.3 | 3 | 522 | NP_033725.2 | |
BC137771.1 | 3 | 322 | ||
BC144897.1 | 3 | 322 | ||
DQ084361.1 | 3 | 260 | AAY86039.1 |
