Assay Details
Target Gene Details
Entrez Gene ID: | 74205 |
Gene Name: | acyl-CoA synthetase long-chain family member 3 |
Gene Aliases: |
2610510B12Rik, Acs3, C85929, Facl3, Pro2194 |
Location: |
Chr.1:78657825-78707743 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 13 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Acsl3 | NM_001033606.2 | NP_001028778.2 | ||
| NM_001136222.1 | NP_001129694.1 | |||
| NM_028817.3 | NP_083093.2 | |||
| AK012088.1 | BAB28022.1 | |||
| AK160839.1 | ||||
| AK161708.1 | BAE36543.1 | |||
| AK166723.1 | ||||
| BC031529.1 | AAH31529.1 |
More Information
Set Membership: |
|

Genomic Map