Assay Details
Target Gene Details
Entrez Gene ID: | 98496 |
Gene Name: | phosphotyrosine interaction domain containing 1 |
Gene Aliases: |
5033414K04Rik, NYGGF4 |
Location: |
Chr.1:84036293-84284675 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Pid1 | NM_001003948.2 | 2 | 416 | NP_001003948.2 |
AK035542.1 | 2 | 291 | ||
AK083256.1 | 2 | 336 | ||
AK150971.1 | 2 | 354 | BAE30002.1 | |
AK151267.1 | 2 | 386 | BAE30254.1 | |
AK164328.1 | 2 | 383 | BAE37740.1 | |
BC054112.1 | 1 | 49 | AAH54112.1 | |
BC080290.1 | 2 | 362 | AAH80290.1 | |
BY004446.1 |
