Assay Details
Target Gene Details
Entrez Gene ID: | 15463 |
Gene Name: | ArfGAP with FG repeats 1 |
Gene Aliases: |
AU045498, C130049H11Rik, C85612, D730048C23Rik, Hrb, RAB, Rip |
Location: |
Chr.1:82839460-82901001 on Build GRCm38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Agfg1 | NM_010472.2 | 11 | 1648 | NP_034602.1 |
| AF057287.1 | 6 | 706 | AAC97477.1 | |
| AK029917.1 | 11 | 1398 | BAC26675.1 | |
| AK048326.1 | 11 | 1649 | BAC33303.1 | |
| AK049400.1 | 10 | 1542 | BAC33736.1 | |
| AK192849.1 | ||||
| AK196110.1 | ||||
| BC031154.1 | 11 | 1635 | AAH31154.1 |

Genomic Map