Assay Details
Target Gene Details
Entrez Gene ID: | 15463 |
Gene Name: | ArfGAP with FG repeats 1 |
Gene Aliases: |
AU045498, C130049H11Rik, C85612, D730048C23Rik, Hrb, RAB, Rip |
Location: |
Chr.1:82839460-82901001 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Agfg1 | NM_010472.2 | 8 | 1304 | NP_034602.1 |
| AF057287.1 | 3 | 362 | AAC97477.1 | |
| AK029917.1 | BAC26675.1 | |||
| AK048326.1 | 8 | 1305 | BAC33303.1 | |
| AK049400.1 | 8 | 1291 | BAC33736.1 | |
| AK198918.1 | 2 | 82 | ||
| BC031154.1 | 8 | 1291 | AAH31154.1 |

Genomic Map