Assay Details
Target Gene Details
Entrez Gene ID: | 213043 |
Gene Name: | aldehyde oxidase 2 |
Gene Aliases: |
Aoh3, Aox3l1 |
Location: |
Chr.1:58276141-58379264 on Build GRCm38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aox2 | NM_001008419.2 | 22 | 2525 | NP_001008419.1 |
| AY187018.2 | 22 | 2525 | AAO38750.2 | |
| AY665589.1 | 22 | 2432 | AAV68256.1 | |
| BC150826.1 | 22 | 2452 |

Genomic Map