Assay Details
Target Gene Details
Entrez Gene ID: | 23797 |
Gene Name: | thymoma viral proto-oncogene 3 |
Gene Aliases: |
AI851531, D930002M15Rik, Nmf350 |
Location: |
Chr.1:177020076-177258289 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Akt3 | NM_011785.3 | NP_035915.3 | ||
| AF124142.1 | AAD29090.1 | |||
| BC066861.1 | AAH66861.1 |
More Information
Set Membership: |
|

Genomic Map