Assay Details
Target Gene Details
Entrez Gene ID: | 52231 |
Gene Name: | ankyrin repeat and zinc finger domain containing 1 |
Gene Aliases: |
1300008P06Rik, 2810025E10Rik, D1Ertd161e |
Location: |
Chr.1:75192160-75199387 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 10 - Intron 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankzf1 | NM_026187.5 | NP_080463.3 | ||
| NR_051998.1 | ||||
| AK004948.2 | ||||
| AK012672.1 | BAB28398.1 | |||
| AK012809.1 | 1 | 774 | ||
| AK156255.1 | BAE33643.1 | |||
| AK157405.1 | ||||
| AW536302.1 | ||||
| BC057896.1 | AAH57896.1 | |||
| BC150879.1 |
Target Gene Details
Entrez Gene ID: | 74577 |
Gene Name: | galactosidase, beta 1-like |
Gene Aliases: |
4833408P15Rik |
Location: |
Chr.1:75198235-75210816 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Glb1l | NM_029010.1 | 17 | 3319 | NP_083286.1 |
| AK029925.1 | 18 | 2803 | BAC26681.1 | |
| AK178874.1 | ||||
| AK180199.1 |
More Information
Set Membership: |
|

Genomic Map