Assay Details
Target Gene Details
Entrez Gene ID: | 52231 |
Gene Name: | ankyrin repeat and zinc finger domain containing 1 |
Gene Aliases: |
1300008P06Rik, 2810025E10Rik, D1Ertd161e |
Location: |
Chr.1:75192160-75199387 on Build GRCm38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ankzf1 | NM_001267620.1 | 4 | 824 | NP_001254549.1 |
| NM_026187.5 | 4 | 824 | NP_080463.3 | |
| NR_051998.1 | 4 | 824 | ||
| NR_051999.1 | 3 | 717 | ||
| AI425962.1 | ||||
| AK004948.2 | 3 | 911 | ||
| AK084573.1 | 3 | 679 | ||
| AK156255.1 | 4 | 788 | BAE33643.1 | |
| AK157405.1 | 4 | 788 | ||
| BC051449.1 | 4 | 804 | AAH51449.1 | |
| BC057896.1 | AAH57896.1 | |||
| BC150879.1 | 4 | 774 | ||
| BU054541.1 | 3 | 225 | ||
| DV062487.1 |

Genomic Map