Assay Details
Target Gene Details
Entrez Gene ID: | 210933 |
Gene Name: | adhesion G protein-coupled receptor B3 |
Gene Aliases: |
A830096D10Rik, Bai3 |
Location: |
Chr.1:25067476-25830206 on Build GRCm38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adgrb3 | NM_175642.4 | 25 | 3588 | NP_783573.4 |
| AK044160.1 | 9 | 1347 | ||
| AK044248.1 | 10 | 1022 | BAC31839.1 | |
| AK081421.1 | 4 | 930 | BAC38215.1 | |
| AY168406.1 | 24 | 3327 | AAO27431.1 | |
| BC032251.1 | 18 | 1805 | AAH32251.1 | |
| BC079670.1 | 23 | 2607 | AAH79670.1 | |
| BC099951.1 | 25 | 3586 | AAH99951.1 |
More Information
Set Membership: |
|

Genomic Map