Assay Details
Target Gene Details
Entrez Gene ID: | 12370 |
Gene Name: | caspase 8 |
Gene Aliases: |
CASP-8, FLICE, MACH, Mch5 |
Location: |
Chr.1:58795229-58847503 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Casp8 | NM_001080126.1 | NP_001073595.1 | ||
| NM_001277926.1 | NP_001264855.1 | |||
| NM_009812.2 | NP_033942.1 | |||
| AF067834.1 | AAC40131.1 | |||
| AJ000641.1 | CAA04196.1 | |||
| AJ007749.1 | CAA07677.1 | |||
| AK146758.1 | BAE27412.1 | |||
| AK153340.1 | BAE31918.1 | |||
| AK156135.1 | ||||
| AK168812.1 | BAE40639.1 | |||
| BC006737.1 | AAH06737.1 | |||
| BC049955.1 | AAH49955.1 | |||
| CT010166.1 | CAJ18374.1 |
More Information
Set Membership: |
|

Genomic Map