Assay Details
Target Gene Details
Entrez Gene ID: | 12168 |
Gene Name: | bone morphogenetic protein receptor, type II (serine/threonine kinase) |
Gene Aliases: |
2610024H22Rik, AL117858, AW546137, BB189135, BMP-2, BMPR-2, BMPR-II, BMPRII, BRK-3, Gm20272 |
Location: |
Chr.1:59764270-59878081 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bmpr2 | NM_007561.4 | NP_031587.1 | ||
| AF003942.1 | AAB63042.1 | |||
| AK148180.1 | BAE28400.1 | |||
| U78048.1 | AAB87638.1 |
More Information
Set Membership: |
|

Genomic Map